Lanthanum(III) phosphate hydrate ≥99.99% (REO, rare earth oxide basis)
Dodavatel: Thermo Scientific
41990.06
41990.14
41990.06EA
1720
CZK
41990.06
41990.14
Lanthanum(III) phosphate hydrate ≥99.99% (REO, rare earth oxide basis)
Lanthanum(III) phosphate hydrate
Purity: 99.99%
CAS No.: 14913-14-5
Molecular Formula: LaPO4.xH2O
Formula Weight: 233.88(anhy)
Physical Form: Lump
Appearance: White to off-white
Odour: Odourless
MDL No.: MFCD00150248
Vzorec:
LaPO₄.xH₂O MW: 233,877 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150248 CAS číslo: 14913-14-5 EINECS: 237-419-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...