jod 99,8, crystals ACS, resublimed
Dodavatel: Thermo Fisher Scientific
|
Varování
|
41955.22
41955.36
41955.22EA
2860
CZK
41955.22
41955.36
jod 99,8, crystals ACS, resublimed
jod
Purity: 99.8%
CAS No.: 7553-56-2
Molecular Formula: I2
Formula Weight: 253.81
Physical Form: Prills or crystals
Density: 4.930
Odour: Irritating
Melting Point: 183-186° subl.
UN No.: UN3495
MDL No.: MFCD00011355
Vzorec:
I₂ MW: 253.81 Bod varu: 184.3 Bod tání: 183…186 °C (subl.) Hustota: 4.930 Storage Temperature: Ambient |
MDL Number:
MFCD00011355 CAS číslo: 7553-56-2 EINECS: 231-442-4 UN: 3495 ADR: 8 III Merck Index: 13,05036 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...