Tisk…

Nickel Aluminium ≥99.0% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

22890.09 22890.18 22890.30
22890.09EA 1980 CZK
22890.09 22890.18 22890.30
Nickel Aluminium ≥99.0% (metals basis)
Nickel Aluminium

Nickel aluminum is used in strengthening constituent in high-temperature nickel-base superalloys, coating blades in gas turbines and jet engines


Purity: 99.0% min CAS No.: 12003-78-0 Molecular Formula: NiAl Physical Form: -100+325 Mesh Powder, Intermetallic Odour: Odourless

Vzorec: NiAl
Storage Temperature: Ambient
MDL Number: MFCD00167392
CAS číslo: 12003-78-0
EINECS: 234-439-6
UN: 3089
ADR: 4.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR