Tisk…

Strontium tungsten oxide ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

41899.14 41899.22
41899.14EA 3190 CZK
41899.14 41899.22
Strontium tungsten oxide ≥99.9% (metals basis)
Strontium tungsten oxide

Purity: 99.9% CAS No.: 13451-05-3 Molecular Formula: O4SrW Formula Weight: 335.46 Physical Form: -200 Mesh Powder Density: 6.187 Appearance: White Odour: Odourless MDL No.: MFCD00049562

Vzorec: O₄SrW
Hustota: 6,187
Storage Temperature: Ambient
MDL Number: MFCD00049562
CAS číslo: 13451-05-3
EINECS: 236-617-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR