Tisk…

Zinc acrylate

Dodavatel: Thermo Fisher Scientific

41780.18 41780.30 41780.A1
41780.18EA 1240 CZK
41780.18 41780.30 41780.A1
Zinc acrylate
Zinc acrylate

CAS No.: 14643-87-9 Molecular Formula: Zn(H2C=CHCO2)2 Formula Weight: 207.50 Physical Form: Powder Density: 1.6 Melting Point: ≈250° UN No.: UN3077 MDL No.: MFCD00067208

Vzorec: Zn(H₂C=CHCO₂)₂
MDL Number: MFCD00067208
CAS číslo: 14643-87-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR