Potassium aquapentachlororuthenate(III) (≥26,4% Ru) ≥99.99% (metals basis), Premion®
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Dipotassium aquapentachlororuthenate(III), Potassium pentachlororuthenate(III) hydrate
41716.03
41716.06
41716.03EA
4430
CZK
41716.03
41716.06
Potassium aquapentachlororuthenate(III) (≥26,4% Ru) ≥99.99% (metals basis), Premion®
Potassium aquapentachlororuthenate(III)
Purity: 99.99%
CAS No.: 14404-33-2
Molecular Formula: K2(RuCl5)(H2O)
Formula Weight: 374.55 (356.53anhy)
Physical Form: Crystalline
Appearance: Brown to red
MDL No.: MFCD00058747
Vzorec:
K₂(RuCl₅)(H₂O) MW: 374,55 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00058747 CAS číslo: 14404-33-2 EINECS: 238-374-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...