Styrene-4-sulphonic acid sodium salt
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Sodium 4-vinylbenzenesulphonate, 4-Vinylbenzenesulphonic acid sodium salt, Sodium-p-styrenesulphonate, p-Styrenesulphonic acid sodium salt
41690.18
41690.30
41690.18EA
1850
CZK
41690.18
41690.30
Styrene-4-sulphonic acid sodium salt
Styrene-4-sulphonic acid sodium salt
CAS No.: 2695-37-6
Molecular Formula: C8H7NaO3S
Formula Weight: 206.19
Physical Form: Powder
Appearance: White
Beilstein Reference: 3575931
MDL No.: MFCD00013379
Vzorec:
C₈H₇NaO₃S MW: 206,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013379 CAS číslo: 2695-37-6 EINECS: 220-266-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...