Poly(styrene sulphonic acid)sodium salt, M.W. 70,000
Dodavatel: Thermo Fisher Scientific
Synonyms:
Poly(sodium-p-styrenesulphonate)
41688.18
41688.30
41688.18EA
2030
CZK
41688.18
41688.30
Poly(styrene sulphonic acid)sodium salt, M.W. 70,000
Poly(styrene sulphonic acid)sodium salt
CAS No.: 25704-18-1
Molecular Formula: [-CH2CH(C6H4)SO3Na)-]n
Physical Form: Powder
MDL No.: MFCD00084449
Vzorec:
[-CH₂CH(C₆H₄)SO₃Na)-]n Storage Temperature: Ambient |
MDL Number:
MFCD00084449 CAS číslo: 25704-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...