Bismuth titanium oxide ≥99.9% (metals basis)
Dodavatel: Thermo Scientific
41669.18
41669.30
41669.18EA
3390
CZK
41669.18
41669.30
Bismuth titanium oxide ≥99.9% (metals basis)
Bismuth titanium oxide
Purity: 99.9%
CAS No.: 12233-34-0
Molecular Formula: Bi2Ti4O11
Formula Weight: 785.42
Physical Form: -325 Mesh Powder
Odour: Odourless
MDL No.: MFCD01310393
Vzorec:
Bi₁₂TiO₂₀ MW: 785,42 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01310393 CAS číslo: 12233-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...