Copper(II) methacrylate, tech.
Dodavatel: Thermo Fisher Scientific
|
Varování
|
41621.06
41621.14
41621.06EA
2000
CZK
41621.06
41621.14
Copper(II) methacrylate, tech.
Copper(II) methacrylate
CAS No.: 19662-59-0
Molecular Formula: C8H10CuO4
Formula Weight: 233.71
Physical Form: Powder
Appearance: Dark blue-green
MDL No.: MFCD00080547
Vzorec:
C₈H₁₀CuO₄ MW: 233.71 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00080547 CAS číslo: 19662-59-0 EINECS: 243-211-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...