Copper(II) ethylacetoacetate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
41489.04
41489.09
41489.18
41489.30
41489.04EA
1190
CZK
41489.04
41489.09
41489.18
41489.30
Copper(II) ethylacetoacetate ≥97%
Copper(II) ethylacetoacetate
Purity: 97%
CAS No.: 14284-06-1
Molecular Formula: (CH3CH2CO2CHCOCH3)2Cu
Formula Weight: 321.81
Physical Form: Powder
Odour: Odourless
Melting Point: 192°
Solubility: Soluble in ethanol
MDL No.: MFCD00017759
Vzorec:
(CH₃CH₂CO₂CHCOCH₃)₂Cu Bod tání: 192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017759 CAS číslo: 14284-06-1 EINECS: 238-181-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...