Molybdenum(V) isopropoxide ≥99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
41484.04
41484.02
41484.04EA
10950
CZK
41484.04
41484.02
Molybdenum(V) isopropoxide ≥99% (metals basis)
Molybdenum(V) isopropoxide
Purity: 99+%
CAS No.: 209733-38-0
Molecular Formula: C15H35MoO5
Formula Weight: 391.04
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Yellow to brown to black
UN No.: UN3181
MDL No.: MFCD00210636
Applications: Precursor used in sol-gel process
Vzorec:
C₁₅H₃₅MoO₅ MW: 391.39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00210636 CAS číslo: 209733-38-0 UN: 3181 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...