1-(tert-Butyldimethylsilyl)imidazole ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
41462.03
41462.06
41462.14
41462.03EA
1970
CZK
41462.03
41462.06
41462.14
1-(tert-Butyldimethylsilyl)imidazole ≥97%
1-(tert-Butyldimethylsilyl)imidazole
Purity: 97%
CAS No.: 54925-64-3
Molecular Formula: C9H18N2Si
Formula Weight: 182.34
Flash Point: 99°(210°F)
Physical Form: Liquid
Density: 0.94
Refractive Index: 1.4805
Sensitivity: Moisture Sensitive
Boiling Point: 247-250°
MDL No.: MFCD00011682
Vzorec:
C₉H₁₈N₂Si Bod varu: 247…250 °C Hustota: 0,94 Bod vzplanutí: 99 °C (210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011682 CAS číslo: 54925-64-3 EINECS: 259-398-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...