Uhličitan stříbrný ∼ 0,7 mmole Ag₂CO₃/g on Celite
Dodavatel: Thermo Fisher Scientific
|
Varování
|
∼0.7 mmole Ag₂CO₃/g reagent
CAS No.: 534-16-7
Molecular Formula: Ag2CO3/Celite
Physical Form: Powder
Sensitivity: Light Sensitive
Odour: Odourless
MDL No.: MFCD00003403
Oxidizing agent for alcohols, diols/triols, hydroquinones, and amines.
Vzorec:
CAg₂O₃ Bod tání: 210…218 °C Hustota: 5,65 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003403 CAS číslo: 534-16-7 EINECS: 208-590-3 UN: 3077 ADR: 9,III Merck Index: 12,08650 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...