Tetramethylammonium hydrogen phthalate ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
41360.14
41360.22
41360.14EA
2070
CZK
41360.14
41360.22
Tetramethylammonium hydrogen phthalate ≥99%
Tetramethylammonium hydrogen phthalate
Purity: 99+%
CAS No.: 79723-02-7
Molecular Formula: C12H17NO4
Formula Weight: 239.27
Physical Form: Crystalline
Melting Point: 148-150°
UN No.: UN2811
MDL No.: MFCD00216634
Applications: In electrolyte formulations
Vzorec:
C₁₂H₁₇NO₄ MW: 239,27 g/mol Bod tání: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216634 CAS číslo: 79723-02-7 EINECS: 416-900-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...