Zinc neodecanoate (17,9 - 18,2% Zn) ≥97%
Dodavatel: Thermo Fisher Scientific
41312.30
41312.A1
41312.A7
41312.30EA
3120
CZK
41312.30
41312.A1
41312.A7
Zinc neodecanoate (17,9 - 18,2% Zn) ≥97%
Zinc neodecanoate
CAS No.: 27253-29-8
Molecular Formula: Zn(C10H19O2)2
Formula Weight: 407.89
Flash Point: 102°(216°F)
Physical Form: Liquid
Appearance: Pale brown
MDL No.: MFCD00014384
Vzorec:
C₂₀H₃₈O₄Zn Bod vzplanutí: 102 °C (216 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014384 CAS číslo: 27253-29-8 EINECS: 248-370-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...