Tisk…

Nickel(II) oxalate hydrate ≥99.9985% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

40757.06 40757.14
40757.06EA 1590 CZK
40757.06 40757.14
Nickel(II) oxalate hydrate ≥99.9985% (metals basis), Puratronic®
Nickel(II) oxalate hydrate

Purity: 99.9985% CAS No.: 126956-48-7 Molecular Formula: C2NiO4.xH2O (x≈2) Formula Weight: 146.73(anhy) Physical Form: Crystalline powder Appearance: Green UN No.: UN3077 MDL No.: MFCD00167155

Vzorec: C₂NiO₄·xH₂O(x≈2)
Storage Temperature: Ambient
MDL Number: MFCD00167155
CAS číslo: 126956-48-7
EINECS: 208-933-7
UN: 3077
ADR: 9,III
Merck Index: 14,6514

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR