Gadolinium(III) hydroxide hydrate ≥99.9% (REO, rare earth oxide basis)
Dodavatel: Thermo Scientific
|
Varování
|
40507.06
40507.14
40507.06EA
2530
CZK
40507.06
40507.14
Gadolinium(III) hydroxide hydrate ≥99.9% (REO, rare earth oxide basis)
Gadolinium(III) hydroxide hydrate
Purity: 99.9%
CAS No.: 100634-91-1
Molecular Formula: Gd(OH)3.xH2O
Formula Weight: 208.27(anhy)
Physical Form: Powder
MDL No.: MFCD00049930
Vzorec:
Gd(OH)₃·xH₂O MW: 208,27 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00049930 CAS číslo: 100634-91-1 EINECS: 240-516-5 Merck Index: 14,4326 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...