Tisk…

Potassium pentachlororhodate(III) (≥28,2% Rh) ≥99.99% (metals basis), Premion®

Dodavatel: Thermo Scientific
Varování

40409.03 40409.06
40409.03EA 25400 CZK
40409.03 40409.06
Potassium pentachlororhodate(III) (≥28,2% Rh) ≥99.99% (metals basis), Premion®
Potassium pentachlororhodate(III)

Purity: 99.99% CAS No.: 65980-75-8 Molecular Formula: K2RhCl5 Formula Weight: 358.37 Physical Form: Powder Solubility: Slightly soluble in cold water. Insoluble in alcohol MDL No.: MFCD00049661

Vzorec: K₂RhCl₅
MW: 358,37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049661
CAS číslo: 65980-75-8
EINECS: 265-989-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR