Tisk…

Cerium triiodide, ultra dry ≥99.9% (REO, rare earth oxide basis)

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Cerium (III) iodide, Cerous iodide

Ampuled under argon

13641.03 13641.06
13641.03EA 3880 CZK
13641.03 13641.06
Cerium triiodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Cerium triiodide
Vzorec: CeI₃
MW: 520,83 g/mol
Bod varu: 1400 °C (1013 hPa)
Bod tání: 766 °C
Storage Temperature: Ambient
MDL Number: MFCD00049423
CAS číslo: 7790-87-6
EINECS: 232-228-3
Merck Index: 14,1999

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR