Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
XSodium ammonium hydrogen phosphate tetrahydrate, Sodium ammonium phosphate, Ammonium sodium phosphate dibasic tetrahydrate, Sodium ammonium phosphate tetrahydrate, Ammonium sodium phosphate tetrahydrate
40320.22
40320.36
40320.A4
40320.22EA
1190
CZK
40320.22
40320.36
40320.A4
Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade
Sodium ammonium hydrogen phosphate tetrahydrate
CAS No.: 7783-13-3
Molecular Formula: NaNH4HPO4.4H2O
Formula Weight: 209.07 (137.02anhy)
Physical Form: Crystalline
Density: 1.544
Odour: Odourless
Melting Point: 79° dec.
Solubility: Soluble in water. Insoluble in alcohol
MDL No.: MFCD00066970
Vzorec:
NaNH₄HPO₄·4H₂O MW: 209,07 g/mol Bod tání: 79 °C (rozkl.) Hustota: 1,544 Storage Temperature: Ambient |
MDL Number:
MFCD00066970 CAS číslo: 7783-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...