Terbium(III) acetylacetonate ≥99.9% (REO, rare earth oxide basis)
Dodavatel: Thermo Fisher Scientific
Synonyms:
Terbium(III) 2,4-pentanedionate
40032.04
40032.09
40032.04EA
3100
CZK
40032.04
40032.09
Terbium(III) acetylacetonate ≥99.9% (REO, rare earth oxide basis)
Terbium(III) acetylacetonate
Purity: 99.9%
CAS No.: 14284-95-8
Molecular Formula: C15H21O6Tb
Formula Weight: 456.25
Physical Form: Powder
Odour: Odourless
MDL No.: MFCD00013503
Vzorec:
C₁₅H₂₁O₆Tb MW: 456.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013503 CAS číslo: 14284-95-8 EINECS: 238-195-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...