Tisk…

Nickel(II) citrate hydrate ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

39634.22 39634.36
39634.22EA 3080 CZK
39634.22 39634.36
Nickel(II) citrate hydrate ≥98%
Nickel(II) citrate hydrate

Purity: 98+% CAS No.: 6018-92-4 Molecular Formula: Ni3(C6H5O7)2.xH2O Formula Weight: 554.33(anhy) Physical Form: Powder UN No.: UN3077 MDL No.: MFCD00054366

Vzorec: Ni₃(C₆H₅O₇)₂·xH₂O
MW: 554,33 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00054366
CAS číslo: 6018-92-4
EINECS: 227-873-2
UN: 3077
ADR: 9,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR