Tisk…

Vanadium(IV) oxide phthalocyanine

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Vanadyl phthalocyanine

39587.02 39587.04 39587.09
39587.02EA 1170 CZK
39587.02 39587.04 39587.09
Vanadium(IV) oxide phthalocyanine
Vanadium(IV) oxide phthalocyanine

CAS No.: 13930-88-6 Molecular Formula: C32H16N8OV Formula Weight: 579.45 Physical Form: Crystalline Appearance: Dark blue to purple UN No.: UN3285 MDL No.: MFCD00041966

Vzorec: C₃₂H₁₆N₈OV
MW: 579.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041966
CAS číslo: 13930-88-6
EINECS: 237-700-2
UN: 3285
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR