Zirconium titanium oxide ≥99.5% (metals basis excluding Hf) (max. 75ppm Hf)
Dodavatel: Thermo Scientific
39490.14
39490.22
39490.14EA
3920
CZK
39490.14
39490.22
Zirconium titanium oxide ≥99.5% (metals basis excluding Hf) (max. 75ppm Hf)
Zirconium titanium oxide
Purity: 99.5%
CAS No.: 12036-70-3
Molecular Formula: ZrTiO4
Formula Weight: 203.11
Physical Form: -325 Mesh Powder
Odour: Odourless
MDL No.: MFCD00210648
Vzorec:
ZrTiO₄ MW: 203,11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00210648 CAS číslo: 12036-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...