Tisk…

Nickel(II) cyanide tetrahydrate

Dodavatel: Thermo Fisher Scientific
Nebezpečí

39480.09 39480.18
39480.09EA 5360 CZK
39480.09 39480.18
Nickel(II) cyanide tetrahydrate
Nickel(II) cyanide tetrahydrate

CAS No.: 13477-95-7 Molecular Formula: Ni(CN)2.4H2O Formula Weight: 182.73 (110.73anhy) Physical Form: -100 Mesh Powder Melting Point: 200° -4H2O Solubility: Insoluble in water. Soluble in ammonium carbonate, ammonia, alkali cyanides UN No.: UN1653 MDL No.: MFCD00049490


Applications: Metallurgy, nickel-plating

Vzorec: Ni(CN)₂·4H₂O
Bod tání: 200 °C (-4H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00049490
CAS číslo: 13477-95-7
EINECS: 209-160-8
UN: 1653
ADR: 6.1,II
Merck Index: 14,6506

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR