Tisk…

Nickel(II) oxalate dihydrate

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Nickel oxalate dihydrate

39454.18 39454.30
39454.18EA 3100 CZK
39454.18 39454.30
Nickel(II) oxalate dihydrate
Nickel(II) oxalate dihydrate

CAS No.: 6018-94-6 Molecular Formula: NiC2O4.2H2O Formula Weight: 182.76 (146.73anhy) Physical Form: Powder Odour: Odourless Solubility: Soluble in acids. Insoluble in water UN No.: UN3077 MDL No.: MFCD00167155

Vzorec: C₂H₄NiO₆
Storage Temperature: Ambient
MDL Number: MFCD00167155
CAS číslo: 6018-94-6
EINECS: 208-933-7
UN: 3077
ADR: 9,III
Merck Index: 14,6514

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR