Tisk…

Nickel(II) phthalocyanine

Dodavatel: Thermo Scientific
Nebezpečí

39453.06 39453.14
39453.06EA 1980 CZK
39453.06 39453.14
Nickel(II) phthalocyanine
Nickel(II) phthalocyanine

CAS No.: 14055-02-8 Molecular Formula: Ni(C32H16N8) Formula Weight: 571.24 Physical Form: Powder Appearance: Dark blue to purple Melting Point: 300° MDL No.: MFCD00041954

Vzorec: Ni(C₃₂H₁₆N₈)
MW: 571,22 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00041954
CAS číslo: 14055-02-8
EINECS: 237-893-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR