Tricarbonyl(cyclooctatetraene)iron ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Cyclooctatetraene iron tricarbonyl, Cyclooctatetraene Iron Tricarbonyl
39402.02
39402.04
39402.02EA
6320
CZK
39402.02
39402.04
Tricarbonyl(cyclooctatetraene)iron ≥98%
Tricarbonyl(cyclooctatetraene)iron
Purity: 98%
CAS No.: 12093-05-9
Molecular Formula: C11H8FeO3
Formula Weight: 244.03
Physical Form: Crystalline
Sensitivity: Air Sensitive
Appearance: Red-brown
Melting Point: 93-95°
UN No.: UN3466
MDL No.: MFCD00001755
Vzorec:
C₁₁H₈FeO₃ MW: 244.03 g/mol Bod tání: 93…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001755 CAS číslo: 12093-05-9 UN: 3466 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...