Tisk…

magnesium pyrophosphate

Dodavatel: Thermo Scientific
Varování

39317.18 39317.30 39317.A1
39317.18EA 2020 CZK
39317.18 39317.30 39317.A1
magnesium pyrophosphate
magnesium pyrophosphate

CAS No.: 13446-24-7 Molecular Formula: Mg2P2O7 Formula Weight: 222.57 Physical Form: Powder Appearance: White MDL No.: MFCD00049482

Vzorec: Mg₂O₇P₂
MW: 222,551 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049482
CAS číslo: 13446-24-7
EINECS: 236-595-0
Merck Index: 14,5684

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR