Acetylacetonatobis(ethylene)rhodium(I) (≥39,9% Rh)
Dodavatel: Thermo Fisher Scientific
39288.03
39288.MD
39288.03EA
34770
CZK
39288.03
39288.MD
Acetylacetonatobis(ethylene)rhodium(I) (≥39,9% Rh)
Acetylacetonatobis(ethylene)rhodium(I)
CAS No.: 12082-47-2
Molecular Formula: C9H15O2Rh
Formula Weight: 258.13
Physical Form: Crystalline
Sensitivity: Air Sensitive
Solubility: Soluble in dichloromethane, chloroform
MDL No.: MFCD00015354
Vzorec:
C₉H₁₆O₂Rh Storage Temperature: Ambient |
MDL Number:
MFCD00015354 CAS číslo: 12082-47-2 EINECS: 235-147-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...