Cerium(III) phosphate ≥99%
Dodavatel: Thermo Fisher Scientific
39215.14
39215.22
39215.36
39215.14EA
1890
CZK
39215.14
39215.22
39215.36
Cerium(III) phosphate ≥99%
Cerium(III) phosphate
Purity: 99% min
CAS No.: 13454-71-2
Molecular Formula: CeO4P
Formula Weight: 235.09
Physical Form: Powder
Density: 5.22
Appearance: Dark yellow
Odour: Odourless
MDL No.: MFCD00049424
Vzorec:
CeO₄P Hustota: 5,22 Storage Temperature: Ambient |
MDL Number:
MFCD00049424 CAS číslo: 13454-71-2 EINECS: 236-637-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...