Tisk…

Lithium niobium methoxide 5% (w/v) in methanol , 99% (metals basis)

Dodavatel: Thermo Scientific
Nebezpečí

39154.AC 39154.AE
39154.ACEA 3570 CZK
39154.AC 39154.AE
Lithium niobium methoxide 5% (w/v) in methanol , 99% (metals basis)
Lithium niobium methoxide

Purity: 99+% CAS No.: 21864-11-9 Molecular Formula: C6H18LiNbO6 Formula Weight: 285.92 Physical Form: Liquid Sensitivity: Moisture Sensitive UN No.: UN1230 MDL No.: MFCD00210627

Vzorec: C₆H₁₈LiNbO₆
MW: 285,92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210627
CAS číslo: 21864-11-9
UN: 1230
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR