Barium niobium oxide ≥99.9% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
39130.09
39130.18
39130.09EA
2320
CZK
39130.09
39130.18
Barium niobium oxide ≥99.9% (metals basis)
Barium niobium oxide
Purity: 99.9%
CAS No.: 12009-14-2
Molecular Formula: BaNb2O6
Formula Weight: 419.11
Physical Form: -100 Mesh Powder
Solubility: Insoluble in water
UN No.: UN1564
MDL No.: MFCD00064751
Vzorec:
BaNb₂O₆ MW: 419.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064751 CAS číslo: 12009-14-2 EINECS: 234-543-1 UN: 1564 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...