Tisk…

Tropaeolin O

Dodavatel: Thermo Scientific

Synonyms: Sodium 2,4-Dihydroxyazobenzene-4'-sulfonate, 2,4-Dihydroxyazobenzene-4'-sulfonic Acid Sodium Salt, Acid Orange 6, Resorcinol Yellow

38708.09 38708.04
38708.09EA 1760 CZK
38708.09 38708.04
Tropaeolin O
Tropaeolin O

CAS No.: 547-57-9 Molecular Formula: C12H9N2NaO5S Formula Weight: 316.27 Physical Form: Powder Appearance: Orange to brown Solubility: Soluble in water and alcohol MDL No.: MFCD00007499

Vzorec: C₁₂H₉N₂O₅SNa
MW: 316,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007499
CAS číslo: 547-57-9
EINECS: 208-924-8
Merck Index: 14,9775

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR