Magnesium(II) phthalocyanine ≥90%, tech.
Dodavatel: Thermo Fisher Scientific
Synonyms:
Magnesium phthalocyanine, Phthalocyanine Magnesium(II)
38601.03
38601.06
38601.03EA
1860
CZK
38601.03
38601.06
Magnesium(II) phthalocyanine ≥90%, tech.
Magnesium(II) phthalocyanine
Purity: 90%
CAS No.: 1661-03-6
Molecular Formula: C32H16MgN8
Formula Weight: 536.84
Physical Form: Powder
MDL No.: MFCD00041953
Vzorec:
C₃₂H₁₆MgN₈ MW: 536.84 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00041953 CAS číslo: 1661-03-6 EINECS: 216-766-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...