Tisk…
Paladium 20% on activated carbon powder standard wetted with ca. 50% water, reduced
Dodavatel: Thermo Fisher Scientific
38308.04 38308.09 38308.18 38308.MD
38308.04EA 4530 CZK
38308.04 38308.09 38308.18 38308.MD
Paladium 20% on activated carbon powder standard wetted with ca. 50% water, reduced
Palladium
Physical Form: 900-1100 m²/g Odour: Odourless
Informace pro objednávání: Sold on a dry weight basis. Unit weight excludes water weight
Vzorec: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS číslo: 7440-05-3