Tisk…
Paladium 10% on activated carbon powder wetted with ca. 50% water for use as a catalyst, unreduced
Dodavatel: Thermo Fisher Scientific
38305.06 38305.14 38305.22
38305.06EA 5670 CZK
38305.06 38305.14 38305.22
Paladium 10% on activated carbon powder wetted with ca. 50% water for use as a catalyst, unreduced
Palladium
Physical Form: 900-1100 m²/g
Vzorec: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS číslo: 7440-05-3