Tisk…

Calcium phosphate tribasic (34 - 40% Ca)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Hydroxylapatite, tri-Calcium phosphate, HAp, Calcium Hydroxide Phosphate, Calcium hydroxyphosphate, Tribasic calcium phosphate

36731.36 36731.A3 36731.A9
36731.36EA 2050 CZK
36731.36 36731.A3 36731.A9
Calcium phosphate tribasic (34 - 40% Ca)
Calcium phosphate tribasic

CAS No.: 12167-74-7 Molecular Formula: Ca10(OH)2(PO4)6 (approximate) Formula Weight: 1004.67 Physical Form: Powder Odour: Odourless MDL No.: MFCD00010904

Vzorec: Ca₅HO₁₃P₃
MW: 502.306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010904
CAS číslo: 12167-74-7
EINECS: 235-330-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR