Titanium(III) chloride aluminum(III) chloride (76.0 - 78.5% TiCl₃)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
36729.22
36729.36
36729.22EA
3950
CZK
36729.22
36729.36
Titanium(III) chloride aluminum(III) chloride (76.0 - 78.5% TiCl₃)
Titanium(III) chloride aluminum(III) chloride
CAS No.: 12003-13-3
Molecular Formula: (TiCl3)3AlCl3
Formula Weight: 596.15
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
UN No.: UN3260
MDL No.: MFCD00145412
Vzorec:
(TiCl₃)₃AlCl₃ MW: 596,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00145412 CAS číslo: 12003-13-3 EINECS: 234-421-8 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...