Iron nickel ≥99.5% (metals basis), powder -325 mesh
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
36683.22
36683.36
36683.14
36683.22EA
3140
CZK
36683.22
36683.36
36683.14
Iron nickel ≥99.5% (metals basis), powder -325 mesh
Iron nickel
Purity: 99.5+%
Molecular Formula: Ni:Fe; 50:50 wt%
Physical Form: Powder
Appearance: Grey
Odour: Odourless
UN No.: UN3089
MDL No.: MFCD01741928
Applications: Low expansion alloy for precision instruments, measuring tapes, weights, etc
Vzorec:
Ni:Fe;₅₀:₅₀% Storage Temperature: Ambient |
MDL Number:
MFCD01741928 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...