Tisk…

Gallium(III) isopropoxide mixture of oligomers ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

36583.03 36583.06
36583.03EA 3540 CZK
36583.03 36583.06
Gallium(III) isopropoxide mixture of oligomers ≥99%
Gallium(III) isopropoxide

Purity: 99% CAS No.: 4452-61-3 Molecular Formula: C9H21GaO3 Formula Weight: 246.98 Flash Point: 68°(154°F) Physical Form: Liquid Density: 1.35 Sensitivity: Moisture Sensitive Appearance: Yellow Melting Point: 72° Boiling Point: dec. UN No.: UN1993 MDL No.: MFCD00144389

Vzorec: C₉H₂₁GaO₃
Bod varu: 120 °C (1 mmHg)
Bod tání: 72 °C
Hustota: 1,35
Bod vzplanutí: 68 °C (154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00144389
CAS číslo: 4452-61-3
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR