Phenylazoformic acid 2-phenylhydrazide compound with 1,5-diphenylcarbohydrazide ACS
Dodavatel: Thermo Fisher Scientific
|
Varování
|
36442.09
36442.18
36442.09EA
1920
CZK
36442.09
36442.18
Phenylazoformic acid 2-phenylhydrazide compound with 1,5-diphenylcarbohydrazide ACS
Phenylazoformic acid 2-phenylhydrazide compound with 1,5-diphenylcarbohydrazide
CAS No.: 10329-15-4
Molecular Formula: C6H5N=NCONHNHC6H5.C6H5NHNHCONHNHC6H5
Formula Weight: 482.55
Physical Form: Powder
Appearance: Orange
Odour: Odourless
Melting Point: 154-156°
Beilstein Reference: 6110063
MDL No.: MFCD00013218
Vzorec:
C₂₆H₂₆N₈O₂ Bod tání: 154…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013218 CAS číslo: 10329-15-4 Merck Index: 14,3322 |