Tisk…

Indium tin oxide ≥99.99% (metals basis)

Dodavatel: Thermo Scientific
Varování

Synonyms: ITO

36348.06 36348.14 36348.22
36348.06EA 2840 CZK
36348.06 36348.14 36348.22
Indium tin oxide ≥99.99% (metals basis)
Indium tin oxide

Purity: 99.99% CAS No.: 50926-11-9 Molecular Formula: In2O3:SnO2; 90:10 wt% Physical Form: -325+500 Mesh Powder Odour: Odourless MDL No.: MFCD00171662

Vzorec: In₂O₃·(SnO₂)x
Storage Temperature: Ambient
MDL Number: MFCD00171662
CAS číslo: 50926-11-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR