Tisk…
2-Hydroxy-2-propanesulphonic acid monosodium salt ≥97%
Dodavatel: Thermo Fisher Scientific
36256.22 36256.36
36256.22EA 3510 CZK
36256.22 36256.36
2-Hydroxy-2-propanesulphonic acid monosodium salt ≥97%
2-Hydroxy-2-propanesulphonic acid monosodium salt
Purity: 97% CAS No.: 540-92-1 Molecular Formula: C3H7NaO4S Formula Weight: 162.14 Physical Form: Crystalline MDL No.: MFCD00031395
Vzorec: C₃H₇NaO₄S
Storage Temperature: Ambient
MDL Number: MFCD00031395
CAS číslo: 540-92-1
EINECS: 208-761-2