Magnesium trisilicate
Dodavatel: Thermo Fisher Scientific
33338.30
33338.A1
33338.30EA
2150
CZK
33338.30
33338.A1
Magnesium trisilicate
Magnesium trisilicate
CAS No.: 14987-04-3
Molecular Formula: Mg2O8Si3
Formula Weight: 260.86
Physical Form: Powder
Appearance: White
Odour: Odourless
MDL No.: MFCD00016203
Vzorec:
Mg₂O₈Si₃ Storage Temperature: Ambient |
MDL Number:
MFCD00016203 CAS číslo: 14987-04-3 EINECS: 239-076-7 Merck Index: 14,5687 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...