Tisk…

Silicic acid hydrate, Reagent Grade

Dodavatel: Thermo Fisher Scientific
Varování

33270.22 33270.36
33270.22EA 2970 CZK
33270.22 33270.36
Silicic acid hydrate, Reagent Grade
Silicic acid hydrate

Silicic acid hydrate is used for studying neurodegenerative diseases due to its ability to inhibit aluminum uptake. It is utilized for the manufacture catalyst and catalyst carrier. It is involved in tungsten filament production as chemical reagent and flux. Further, it is used for oil and wax decoloring.


CAS No.: 1343-98-2 Molecular Formula: SiO2.xH2O Formula Weight: 60.09(anhy) Physical Form: -100 Mesh Powder Appearance: White MDL No.: MFCD00054122

Vzorec: SiO₂·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00054122
CAS číslo: 1343-98-2
EINECS: 215-683-2
Merck Index: 14,8490

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR