Zinc protoporphyrin ≥96%
Dodavatel: Thermo Scientific
Synonyms:
Zinc(II) 3,3'-(3,8,13,17-tetramethyl-7,12-divinylporphyrin-2,18-diyl)dipropionate, Protoporphyrin IX zinc(II), Zinc (II) Protoporphyrin IX
32068.MA
32068.MF
32068.01
32068.MAEA
2120
CZK
32068.MA
32068.MF
32068.01
Zinc protoporphyrin ≥96%
Zinc protoporphyrin
Purity: 96%
CAS No.: 15442-64-5
Molecular Formula: C34H32N4O4Zn
Formula Weight: 626.04
Physical Form: Crystalline
Appearance: Dark red to purple
Odour: Odourless
MDL No.: MFCD00011612
Vzorec:
C₃₄H₃₂N₄O₄Zn MW: 626,034 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011612 CAS číslo: 15442-64-5 EINECS: 239-455-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...