Oxid hlinitý aktivovaný neutrální Brockmann Grade I 58 angstroms
Dodavatel: Thermo Fisher Scientific
11502.A7
11502.30
11502.A7EA
10640
CZK
11502.A7
11502.30
Oxid hlinitý aktivovaný neutrální Brockmann Grade I 58 angstroms
Aluminium oxide activated neutral
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: -60 Mesh Powder, S.A. 150m2/g
Odour: Odourless
Melting Point: 2045°
MDL No.: MFCD00003424
Suggested for the separation of certain steroids, alkaloids, hydrocarbons, esters, aldehydes, alcohols and weak organic acids and bases. It is usually used with non-aqueous solutions
Vzorec:
Al₂O₃ MW: 101.96 g/mol Hustota: 3.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS číslo: 1344-28-1 EINECS: 215-691-6 REACH: 01-2119529248-35 Merck Index: 14,356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...