Tisk…
Ceramic tubes, both ends open
Dodavatel: Thermo Fisher Scientific
31606.EH 31606.EO 31609.EH
31606.EHEA 9510 CZK
31606.EH 31606.EO 31609.EH
Ceramic tubes, both ends open
Hadičky Přenosová hadička
Pure Al₂O₃ Aluminium oxide AL24.
Slightly porous, good resistance to thermal shock, extremely good creep strength.